Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAGPRGALLA WCRRQCEGYR GVEIRDLSSS FRDGLAFCAI LHRHRPDLLD FDSLSKDNVF ENNRLAFEVA EKELGIPALL DPNDMVSMSV PDCLSIMTYV SQYYNHFCSP GQAGVSPPRK GLAPCSPPSV APTPVEPEDV AQGEELSSGS LSEQGTGQTP SSTCAACQQH VHLVQRYLAD GRLYHRHCFR CRRCSSTLLP GAYENGPEEG TFVCAEHCAR LGPGTRSGTR PGPFSQPKQQ HQQQLAEDAK DVPGGGPSSS APAGAEADGP KASPEARPQI PTKPRVPGKL QELASPPAGR PTPAPRKASE STTPAPPTPR PRSSLQQENL VEQAGSSSLV NGRLHELPVP KPRGTPKPSE GTPAPRKDPP WITLVQAEPK KKPAPLPPSS SPGPPSQDSR QVENGGTEEV AQPSPTASLE SKPYNPFEEE EEDKEEEAPA APSLATSPAL GHPESTPKSL HPWYGITPTS SPKTKKRPAP RAPSASPLAL HASRLSHSEP PSATPSPALS VESLSSESAS QTAGAELLEP PAVPKSSSEP AVHAPGTPGN PVSLSTNSSL ASSGELVEPR VEQMPQASPG LAPRTRGSSG PQPAKPCSGA TPTPLLLVGD RSPVPSPGSS SPQLQVKSSC KENPFNRKPS PAASPATKKA TKGSKPVRPP APGHGFPLIK RKVQADQYIP EEDIHGEMDT IERRLDALEH RGVLLEEKLR GGLNEGREDD MLVDWFKLIH EKHLLVRRES ELIYVFKQQN LEQRQADVEY ELRCLLNKPE KDWTEEDRAR EKVLMQELVT LIEQRNAIIN CLDEDRQREE EEDKMLEAMI KKKEFQREAE PEGKKKGKFK TMKMLKLLGN KRDAKSKSPR DKS
MICALL1 may be a cytoskeletal regulator.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: L23MRP; L23mt; MICAL-like 1; MICAL-like protein 1; MIRab13; Molecule interacting with Rab13; MRP-L23; RPL23; RPL23L
Gene Aliases: KIAA1668; MICAL-L1; MICALL1; MIRAB13
UniProt ID: (Human) Q8N3F8
Entrez Gene ID: (Human) 85377
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support