Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: SLVGSNVYI TPAGSQGLPP HYDDVEVFIL QLEGEKHWRL YHPTVPLARE YSVEAEERIG RPVHEFMLKP GDLLYFPRGT IHQADTPAGL AHSTHVTIST YQNNSWGDFL LDTISGLVFD TAKEDVELRT GIPRQLLLQV ESTTVATRRL SGFLRTLADR LEGTKELLSS DMKKDFIMHR LPPYSAGDGA ELSTPGGKLP RLDSVVRLQF KDHIVLTVLP DQDQSDETQE KMVYIYHSLK NSRETHMMGN EEETEFHGLR FPLSHLDALK QIWNSPAISV KDLKLTTDEE KESLVLSLWT ECLIQVV (160-465 aa encoded by BC014928 )
MINA is nuclear localized, myc-inducible protein that is thought to play a role in mammalian cell proliferation. Treatment of cancer cells lines such as the colon cancer cell line SW680 with siRNA against MINA inhibits cell growth, demonstrating that MINA may be a potential therapeutic target. MINA regulates several genes related to cell adhesion and metabolism that have also been shown to be regulated by c-Myc, but also regulates other genes whose expression are not modulated by c-Myc such as EGFR, IL-6 and HGF. MINA has also been found to act as a repressor to IL-4 expression in T cells, indicating that it may also play a role in T cell differentiation and genetic variation in T helper type 2 bias.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 60S ribosomal protein L27a histidine hydroxylase; Bifunctional lysine-specific demethylase and histidyl-hydroxylase MINA; DKFZp762O1912; FLJ14393; Histone lysine demethylase MINA; MINA; mina {ECO:0000312|RGD:708521}; Mina-53; mineral dust induced gene protein; Mineral dust-induced gene protein; MYC-induced nuclear antigen; myc-induced nuclear antigen, 53 kDa; Nucleolar protein 52; Protein mina53; Ribosomal oxygenase 2; Ribosomal oxygenase MINA; ROX
Gene Aliases: 1810047J07Rik; 2410057H13Rik; 3830408E23Rik; AI449204; MDIG; MINA; MINA53; NO52; RIOX2; ROX
UniProt ID: (Human) Q8IUF8, (Mouse) Q8CD15, (Rat) Q8CFC1
Entrez Gene ID: (Human) 84864, (Mouse) 67014, (Rat) 266670
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support