Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The protein encoded by this gene interacts with the transcription factor myocardin, a key regulator of smooth muscle cell differentiation. The encoded protein is predominantly nuclear and may help transduce signals from the cytoskeleton to the nucleus. This gene is involved in a specific translocation event that creates a fusion of this gene and the RNA-binding motif protein-15 gene. This translocation has been associated with acute megakaryocytic leukemia.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: basic, SAP and coiled-coil domain; Megakaryoblastic leukemia 1 protein; Megakaryocytic acute leukemia protein; mixed lineage leukemia; MKL/myocardin-like protein 1; MRTF-A; Myocardin-related transcription factor A
Gene Aliases: BSAC; KIAA1438; MAL; MKL1; MRTF-A; MRTFA
UniProt ID: (Human) Q969V6
Entrez Gene ID: (Human) 57591
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support