Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Polypeptide reacting: MLDP = myocardial lipid droplet protein (also described as OXPAT/PAT-1 and perilipin 5/PLIN5), MW 50,844 (calculated from aa sequence data); apparent MW 52,000 (after SDS-PAGE). MLDP/PLIN5 pertains to the PLIN/PAT-family proteins, covering the surface of cytoplasmic lipid droplets and sharing a homologous domain called PAT. Additional PLIN/PAT proteins include adipophilin (ADRP/PLIN2), perilipin (PLIN1), and TIP47 (PLIN3) which are expressed in differentiation-related stages of lipid metabolism.MLDP/PLIN5 is positively detected in heart and skeletal muscle.Reactivity on cultured cell lines: Caco-2, PLC.
Lipid droplet-associated protein that maintains the balance between lipogenesis and lipolysis and also regulates fatty acid oxidation in oxidative tissues. Recruits mitochondria to the surface of lipid droplets and is involved in lipid droplet homeostasis by regulating both the storage of fatty acids in the form of triglycerides and the release of fatty acids for mitochondrial fatty acid oxidation. In lipid droplet triacylglycerol hydrolysis, plays a role as a scaffolding protein for three major key lipolytic players: ABHD5, PNPLA2 and LIPE. Reduces the triacylglycerol hydrolase activity of PNPLA2 by recruiting and sequestering PNPLA2 to lipid droplets. Phosphorylation by PKA enables lipolysis probably by promoting release of ABHD5 from the perilipin scaffold and by facilitating interaction of ABHD5 with PNPLA2. Also increases lipolysis through interaction with LIPE and upon PKA-mediated phosphorylation of LIPE.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Lipid storage droplet protein 5; Perilipin-5
Gene Aliases: LSDA5; LSDP5; MLDP; OXPAT; PAT-1; PLIN5
UniProt ID: (Human) Q00G26
Entrez Gene ID: (Human) 440503
Molecular Function:
scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support