Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: QVLQQVDNK FIACLMSTKT EENGEADSYE KQQAQGSGRK KLLSSTLIPP LEITVTEEQR RLLWCYHKNL EDLGLEFVFP DTSDSLVLVG KVPLCFVERE ANELRRGRST VTKSIVEEFI REQLELLQTT GGIQGTLPLT VQKVLASQAC HGAIKFNDGL SLQESCRLIE ALSSCQLPFQ CAHGRPSMLP LADIDHLEQE KQIKPNLTKL RKMAQAWRLF GKAECDTRQS LQQSMPPCEP P (1190-1430 aa encoded by BC112167)
This gene is a member of the MutL-homolog (MLH) family of DNA mismatch repair (MMR) genes. MLH genes are implicated in maintaining genomic integrity during DNA replication and after meiotic recombination. The protein encoded by this gene functions as a heterodimer with other family members. Somatic mutations in this gene frequently occur in tumors exhibiting microsatellite instability, and germline mutations have been linked to hereditary nonpolyposis colorectal cancer type 7 (HNPCC7). Several alternatively spliced transcript variants have been identified, but the full-length nature of only two transcript variants has been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DNA mismatch repair protein Mlh3; MGC138372; MutL protein homolog 3
Gene Aliases: HNPCC7; MLH3
UniProt ID: (Human) Q9UHC1
Entrez Gene ID: (Human) 27030
Molecular Function: DNA metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support