Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: VKSTAYLSPQ ELEDVFYQYD VKSEIYSFGI VLWEIATGDI PFQGCNSEKI RKLVAVKRQQ EPLGEDCPSE LREIIDECRA HDPSVRPSVD EILKKLSTFS K
The protein kinase domain is predicted to be catalytically inactive. Molecular function: protein binding. There are two isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hMLKL; Mixed lineage kinase domain-like protein; MLKL
Gene Aliases: hMLKL; MLKL
UniProt ID: (Human) Q8NB16
Entrez Gene ID: (Human) 197259
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support