Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • MLL4 Antibodies

            Abnova

            MLL4 Monoclonal Antibody (1F6)

            View all (9) MLL4 antibodies
            Datasheet
            Protocols
            Questions & Answers
            Datasheet
            Protocols
            Questions & Answers

            Cite MLL4 Monoclonal Antibody (1F6)

            • Antibody Testing Data (1)
            Application
            MLL4 Antibody in ELISA (ELISA)
            Group 53 Created with Sketch.

            FIGURE: 1 / 1

            MLL4 Antibody (H00009757-M01) in ELISA

            Detection limit for recombinant GST tagged MLL4 is approximately 0.1 ng/mL as a capture antibody.
            View Product

            MLL4 Antibody in ELISA (ELISA)

            Product Details

            H00009757-M01

            Applications
            Tested Dilution
            Publications

            Western Blot (WB)

            1-5 µg/mL
            -

            ELISA (ELISA)

            0.1 ng/mL
            -
            Product Specifications

            Species Reactivity

            Human

            Host/Isotype

            Mouse / IgG1, kappa

            Class

            Monoclonal

            Type

            Antibody

            Clone

            1F6

            Immunogen

            MLL4 (NP_055542, 813 a.a. approximately 904 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
            View immunogen

            Conjugate

            Unconjugated

            Form

            Liquid

            Concentration

            See Label

            Purification

            Affinity chromatography

            Storage buffer

            PBS, pH 7.4

            Contains

            no preservative

            Storage conditions

            -20°C, Avoid Freeze/Thaw Cycles

            Shipping conditions

            Ambient (domestic); Wet ice (international)

            Product Specific Information

            Sequence of this protein is as follows: KVAASMPLSP GGQMEEVAGA VKQISDRGPV RSEDESVEAK RERPSGPESP VQGPRIKHVC RHAAVALGQA RAMVPEDVPR LSALPLRDRQ DL

            Target Information

            This gene encodes a protein which contains multiple domains including a CXXC zinc finger, three PHD zinc fingers, two FY-rich domains, and a SET domain. The SET domain is a conserved C-terminal domain that characterizes proteins of the MLL family. This gene is ubiquitously expressed in adult tissues. It is also amplified in solid tumor cell lines, and may be involved in human cancer. Two alternatively spliced transcript variants encoding distinct isoforms have been reported for this gene, however, the full length nature of the shorter transcript is not known.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            References (0)

            Have you cited this product in a publication?
            Let us know so we can reference it here.
            Cite this product

            Bioinformatics

            Protein Aliases: Histone-lysine N-methyltransferase 2B; histone-lysine N-methyltransferase MLL4; KMT2D; lysine (K)-specific methyltransferase 2B; Lysine N-methyltransferase 2B; mixed lineage leukemia gene homolog 2; myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila) 4; Myeloid/lymphoid or mixed-lineage leukemia protein 4; Trithorax homolog 2; trithorax homologue 2; WBP-7; WW domain binding protein 7; WW domain-binding protein 7

            View more View less

            Gene Aliases: CXXC10; HRX2; KIAA0304; KMT2B; MLL1B; MLL2; MLL4; TRX2; WBP-7; WBP7

            View more View less

            UniProt ID: (Human) Q9UMN6

            View more View less

            Entrez Gene ID: (Human) 9757

            View more View less

            Function(s)

            DNA binding transcription factor activity, sequence-specific DNA binding protein binding zinc ion binding histone-lysine N-methyltransferase activity histone methyltransferase activity (H3-K4 specific) histone modifying enzyme

            Process(es)

            ovarian follicle development transcription, DNA-templated memory oocyte differentiation gene silencing ovulation chromatin-mediated maintenance of transcription histone H3-K4 methylation regulation of histone H3-K4 methylation histone H3-K4 trimethylation
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            United States flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-56756dfb8b-flzw4:80/100.66.79.199:80.
            git-commit: de6905f249d25e22729e613aef254bb544bd8b7c
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.31.1-2025.06.25-1.0