Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MGKKLDLSKL TDEEAQHVLE VVQRDFDLRR KEEERLEALK GKIKKESSKR ELLSDTAHLN ETHCARCLQP YQLLVNSKRQ CLECGLFTCK SCGRVHPEEQ GWICDPCHLA RVVKIGSLEW YYEHVKARFK RFGSAKVIRS LHGRLQGGAG PELISEERSG DSDQTDEDGE PGSEAQAQAQ PFGSKKKRLL SVHDFDFEGD SDDSTQPQGH SLHLSSVPEA RDSPQSLTDE SCSEKAAPHK AEGLEEADTG ASGCHSHPEE QPTSISPSRH GALAELCPPG GSHRMALGTA AALGSNVIRN EQLPLQYLAD VDTSDEESIR AHVMASHHSK RRGRASSESQ IFELNKHISA VECLLTYLEN TVVPPLAKGL GAGVRTEADV EEEALRRKLE ELTSNVSDQE TSSEEEEAKD EKAEPNRDKS VGPLPQADPE VGTAAHQTNR QEKSPQDPGD PVQYNRTTDE ELSELEDRVA VTASEVQQAE SEVSDIESRI AALRAAGLTV KPSGKPRRKS NLPIFLPRVA GKLGKRPEDP NADPSSEAKA MAVPYLLRRK FSNSLKSQGK DDDSFDRKSV YRGSLTQRNP NARKGMASHT FAKPVVAHQS
Slac2-a (for synaptotagmin-like protein (Slp) homolog lacking C2 domains-a) links Rab27A on melanosomes with Myosin VA in melanocytes. Slac2-a binds Myosin VA through a C-terminal region and GTP-bound Rab27A through its synaptotagmin-like protein homology domain (SHD), which is located near the N-terminus. The transport of pigment and cytotoxic granules in melanocytes requires the stable formation of this complex, and thus mutations in the binding domains of the three protein components may cause albinism and/or severe immune disorders.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Exophilin-3; leaden; Melanophilin; MGC2771; MGC59733; OTTMUSP00000025451; SlaC2-a; Slp homolog lacking C2 domains a; Synaptotagmin-like protein 2a
Gene Aliases: MLPH; SLAC2-A; SLAC2A
UniProt ID: (Human) Q9BV36
Entrez Gene ID: (Human) 79083
Molecular Function: myelin protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support