Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAVCGLGSR LGLGSRLGLR GCFGAARLLY PRFQSRGPQG VEDGDRPQPS SKTPRIPKIY TKTGDKGFSS TFTGERRPKD DQVFEAVGTT DELSSAIGFA LELVTEKGHT FAEELQKIQC TLQDVGSALA TPCSSAREAH LKYTTFKAGP ILELEQWIDK YTSQLPPLTA FILPSGGKIS SALHFCRAVC RRAERRVVPL VQMGETDANV AKFLNRLSDY LFTLARYAAM KEGNQEKIYM KNDPSAESEG L (1-250 aa encoded by BC011831)
This gene encodes a protein that catalyzes the final step in the conversion of vitamin B(12) into adenosylcobalamin (AdoCbl), a vitamin B12-containing coenzyme for methylmalonyl-CoA mutase. Mutations in the gene are the cause of vitamin B12-dependent methylmalonic aciduria linked to the cblB complementation group.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: aquocob(I)alamin vitamin B12s adenosyltransferase; ATP:co(I)rrinoid adenosyltransferase MMAB; ATP:Cob(I)alamin Adenosyltransferase; ATP:corrinoid adenosyltransferase; cilia and flagella associated protein 23; cob(I)alamin adenosyltransferase; cob(I)yrinic acid a,c-diamide adenosyltransferase, mitochondrial; Corrinoid adenosyltransferase MMAB; Methylmalonic aciduria type B homolog; Methylmalonic aciduria type B protein
Gene Aliases: 9130222L19Rik; ATR; cblB; CFAP23; cob; MMAB
UniProt ID: (Human) Q96EY8, (Mouse) Q9D273
Entrez Gene ID: (Human) 326625, (Mouse) 77697
Molecular Function: dehydrogenase oxidoreductase metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support