Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Predicted reactivity by homology: Orangutan, Bovine.
Mmgt1 encodes a protein that that mediates Mg(2+) transport.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ER membrane protein complex subunit 5; Membrane magnesium transporter 1; Transmembrane protein 32
Gene Aliases: 9630048L06Rik; BC032271; EMC5; mir-5116; Mir5116; MMGT1; TMEM32
UniProt ID: (Human) Q8N4V1, (Mouse) Q8K273
Entrez Gene ID: (Human) 93380, (Mouse) 236792
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support