Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteins of the matrix metalloproteinase (MMP) family are involved in the breakdown of extracellular matrix in normal physiological processes, such as embryonic development, reproduction, and tissue remodeling, as well as in disease processes, such as arthritis and metastasis. Most MMP's are secreted as inactive proproteins which are activated when cleaved by extracellular proteinases. This gene encodes a secreted enzyme which breaks down the interstitial collagens, types I, II, and III. The gene is part of a cluster of MMP genes which localize to chromosome 11q22.3. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Collagenase; collagenase-like A; Interstitial collagenase A; matrix metallo protease; matrix metallopeptidase 1a; matrix metallopeptidase 1a (interstitial collagenase); matrix metalloproteinase 1a (interstitial collagenase); Matrix metalloproteinase-1a; Matrix metalloproteinase1; Mcol-A; MMP; MMP-1a; MMPs
Gene Aliases: Mcol-A; McolA; Mmp1a
UniProt ID: (Mouse) Q9EPL5
Entrez Gene ID: (Rat) 300339, (Mouse) 83995
Molecular Function:
metalloprotease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support