Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Positive control: Colorectal cancer
Cellular location: Cell membrane, Secreted.
MMP20 degrades amelogenin, the major protein component of the enamel matrix, as well as two of the macromolecules characterizing the cartilage extracellular matrix: aggrecan and the cartilage oligomeric matrix protein (COMP). MMP20 may play a central role in tooth enamel formation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Enamel metalloproteinase; Enamelysin; matrix metallo protease; matrix metalloproteinase 20 (enamelysin); Matrix metalloproteinase-20; MMP; MMP-20; MMPs
Gene Aliases: AI2A2; MMP-20; MMP20
UniProt ID: (Human) O60882
Entrez Gene ID: (Human) 9313
Molecular Function:
metalloprotease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support