Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Positive control: Human placenta, human testis lysate
Cellular location: Secreted, extracellular space, extracellular matrix.
Epilysin, also known as matrix metalloproteinase-28 (MMP-28), is a secreted protein that belongs to the peptidase M10A family. Matrix metalloproteinase proteins are responsible for the breakdown of the extracellular matrix which is important for normal physiological processes such as tissue remodeling, reproduction and embryonic development. Epilysin is produced by proliferating keratinocytes and is responsible for mediating the degradation of casein. Its expression is upregulated in response to injury of the skin suggesting that Epilysin may be involved in tissue repair and homeostasis. In addition, Epilysin may function predominantly in the nervous system. Incubation of Epilysin with rat embryonic brain tissue results in the degradation of myelin proteins. In Xenopus, Epilysin localizes to nerves and is upregulated during neurulation. This suggests that Epilysin plays an evolutionarily conserved role in neural development and may be involved in modulating the axonal-glial extracellular environment.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Epilysin; matrix metallo protease; matrix metalloprotease MMP25; Matrix metalloproteinase-28; MMP; MMP-28; MMPs; PRO4339; UNQ1893
Gene Aliases: EPILYSIN; MM28; MMP-25; MMP-28; MMP25; MMP28; UNQ1893/PRO4339
UniProt ID: (Human) Q9H239
Entrez Gene ID: (Human) 79148
Molecular Function:
metalloprotease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support