Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: AALVWGPEKN KIYFFRGRDY WRFHPSTRRV DSPVPRRATD WRGVPSEIDA AFQDADGYAY FLRGRLYWKF DPVKVKALEG FPRLVGPDFF GCAEPANTFL
Stromelysin, a member of the matrix metalloproteinase family, demonstrates wide substrate specificity with the ability to degrade proteoglycan, fibronectin, laminin, casein, and the nonhelical region of collagen. The stromelysin-3 (MMP-11) gene was originally identified on the basis that it is expressed specifically in stromal cells surrounding invasive breast carcinomas. MMP-11 is localized by in situ hybridization to the long arm of chromosome 22. The MMP-11 expression is found more specifically in malignant tumors than in benign ones.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: matrix metallo protease; matrix metallopeptidase 11 (stromelysin 3); Matrix metalloproteinase 11 (stromelysin 3); Matrix metalloproteinase-11; MMP; MMP-11; MMPs; SL-3; stromelysin III; Stromelysin-3
Gene Aliases: MMP-11; MMP11; SL-3; ST3; STMY3
UniProt ID: (Human) P24347, (Mouse) Q02853, (Rat) Q499S5
Entrez Gene ID: (Human) 4320, (Mouse) 17385, (Rat) 25481
Molecular Function:
metalloprotease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support