Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Immunogen sequence: ATTGACCTTG GCACCGGCCC CACCCCCACG CTGGGCCCTG TCACTCCTGA GATCTGCAAA CAGGACATTG TATTTGATGG CATCGCTCAG ATCCGTGGTG AGATCTTCTT CTTCAAGGAC CGGTTCATTT GGCGGACTGT GACGCCACGT GACAAGCCCA TGGGGCCCCT GCTGGTGGCC ACATTCTGGC CTGAGCTCCC GGAAAAGATT GATGCGGTAT ACGAGGCCCC ACAGGAGGAG AAGGCTGTGT TCTTTGCAGG GAATGAATAC TGGATCTACT CAGCCAGCAC CCTGGAGCGA GGGTACCCCA AGCCACTGAC CAGCCTGGGA CTGCCCCCTG ATGTCCAGCG AGTGGATGCC GCCTTTAACT GGAGCAAAAA CAAGAAGACA TACATCTTTG CTGGAGACAA ATTCTGGAGA TACAATGAGG TGAAGAAGAA AATGGATCCT GGCTTCCCCA AGCTCATCGC AGATGCCTGG AATGCCATCC CCGATAACCT GGATGCCGTC GTGGACCTGC AGGGCGGCGG TCACAGCTAC TTCTTCAAGG GTGCCTATTA CCTGAAGCTG GAGAACCAAA GTCTGAAGAG CGTGAAGTTT GGAAGCATCA AATCCGACTG GCTAGGCTGC
MMP (matrix metalloproteinase) are proteolytic enzymes capable of degrading connective tissue components. MMP have a common mode of activation, a conserved amino acid sequence in the putative metal binding-active site region, and are inhibited by specific tissue inhibitors of metalloproteinases (TIMPs). MMPa and TIMPs play a significant role in regulating angiogenesis. MMP2 is synthesized as a 631 amino acid proenzyme which is activated by cleavage of the first 80 amino acids, and contains the basic structure of propeptide, catalytic, and hemopexin domains. The matrix metalloproteinases (MMPs) are a family of at least eighteen secreted and membrane-bound zincendopeptidases. Collectively, these enzymes can degrade all the components of the extracellular matrix, including fibrillar and non-fibrillar collagens, fibronectin, laminin and basement membrane glycoproteins. In general, a signal peptide, a propeptide, and a catalytic domain containing the highly conserved zinc-binding site characterizes the structure of the MMPs. Functionally, MMP2 is involved in tissue remodeling. Mutations in MMP-2 gene have been associated with Winchester syndrome and Nodulosis-Arthropathy-Osteolysis (NAO) syndrome. Two transcript variants encoding different isoforms of MMP-2 have been found.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 72 kDa gelatinase; 72 kDa type IV collagenase; 72kD gelatinase; 72kD type IV collagenase; 72kDa gelatinase; 72kDa type IV collagenase; CLG 4A; Collagenase; collagenase type IV-A; Gelatinase A; Gelatinase alpha; Mat; matrix metallo protease; matrix metallopeptidase 2 (gelatinase A, 72kDa gelatinase, 72kDa type IV collagenase); matrix metalloproteinase 2; Matrix metalloproteinase-2; matrix metalloproteinase-II; MMP; MMP-2; MMPs; neutrophil gelatinase; Progelatinase A; TBE-1; TBE1
Gene Aliases: CLG4; CLG4A; GelA; MMP-2; MMP-II; MMP2; MONA; TBE-1
UniProt ID: (Human) P08253, (Rat) P33436, (Mouse) P33434
Entrez Gene ID: (Human) 4313, (Rat) 81686, (Mouse) 17390
Molecular Function:
metalloprotease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support