Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
MMP20 degrades amelogenin, the major protein component of the enamel matrix, as well as two of the macromolecules characterizing the cartilage extracellular matrix: aggrecan and the cartilage oligomeric matrix protein (COMP). MMP20 may play a central role in tooth enamel formation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Enamel metalloproteinase; Enamelysin; matrix metallo protease; matrix metallopeptidase 20 (enamelysin); matrix metalloproteinase 20 (enamelysin); Matrix metalloproteinase-20; MMP; MMP-20; MMPs
Gene Aliases: AI2A2; MMP-20; MMP20
UniProt ID: (Human) O60882, (Mouse) P57748
Entrez Gene ID: (Human) 9313, (Mouse) 30800, (Rat) 300341
Molecular Function:
metalloprotease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support