Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • MMP-9 Antibodies

          Proteintech

          MMP9 (N-terminal) Polyclonal Antibody

          1,017 References
          View all (82) MMP-9 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite MMP9 (N-terminal) Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (26)
          MMP9 (N-terminal) Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          MMP9 (N-terminal) Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 26

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          MMP9 (N-terminal) Antibody (10375-2-AP) in ICC/IF

          Immunofluorescent analysis of HeLa cells using 10375-2-AP (MMP9 (N-terminal) antibody) at dilution of 1:50 and Alexa Fluor 488-conjugated AffiniPure Goat Anti-Rabbit IGG (H+L). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          MMP9 (N-terminal) Antibody in Immunocytochemistry (ICC/IF)
          MMP9 (N-terminal) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MMP9 (N-terminal) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MMP9 (N-terminal) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MMP9 (N-terminal) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MMP9 (N-terminal) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MMP9 (N-terminal) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MMP9 (N-terminal) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MMP9 (N-terminal) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MMP9 (N-terminal) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MMP9 (N-terminal) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MMP9 (N-terminal) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MMP9 (N-terminal) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MMP9 (N-terminal) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MMP9 (N-terminal) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MMP9 (N-terminal) Antibody in Western Blot (WB)
          MMP9 (N-terminal) Antibody in Western Blot (WB)
          MMP9 (N-terminal) Antibody in Western Blot (WB)
          MMP9 (N-terminal) Antibody in Western Blot (WB)
          MMP9 (N-terminal) Antibody in Western Blot (WB)
          MMP9 (N-terminal) Antibody in Western Blot (WB)
          MMP9 (N-terminal) Antibody in Western Blot (WB)
          MMP9 (N-terminal) Antibody in Western Blot (WB)
          MMP9 (N-terminal) Antibody in Western Blot (WB)
          MMP9 (N-terminal) Antibody in Flow Cytometry (Flow)
          MMP9 (N-terminal) Antibody in Immunoprecipitation (IP)
          MMP9 (N-terminal) Polyclonal Antibody

          Product Details

          10375-2-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          View 822 publications 822 publications

          Immunohistochemistry (IHC)

          -
          View 126 publications 126 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:500
          -

          Immunocytochemistry (ICC/IF)

          1:20-1:200
          View 52 publications 52 publications

          Flow Cytometry (Flow)

          Assay-dependent
          -

          ELISA (ELISA)

          -
          View 1 publication 1 publication

          Immunoprecipitation (IP)

          0.5-4.0 µg
          View 2 publications 2 publications

          Miscellaneous PubMed (Misc)

          -
          View 4 publications 4 publications

          Not applicable (N/A)

          -
          View 10 publications 10 publications
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Hamster, Human, Mouse, Plant, Rabbit, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          MMP9 (N-terminal) Fusion Protein Ag0552 (1-100 aa encoded by BC006093)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.27 mg/mL

          Amount

          40.5 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: MSLWQPLVL VLLVLGCCFA APRQRQSTLV LFPGDLRTNL TDRQLAEEYL YRYGYTRVAE MRGESKSLGP ALLLLQKQLS LPETGELDSA TLKAMRTPRC G (1-100 aa encoded by BC006093)

          Target Information

          MMP9 (matrix metallopeptidase 9, GELB, CLG4B) is a matrix metalloproteinase, a family of zinc and calcium-dependent endopeptidases that degrade extracellular matrix proteins. MMP9 is secreted as a 92kDa zymogen and cleavage of pro-MMP9 results in the active enzyme with a molecular weight of 82kDa. MMP9 has a gelatin-binding domain consisting of three fibronectin type II units, a catalytic domain containing the zinc-binding site, a proline-rich type V collagen-homologous domain and a hemopexin-like domain. MMP9 is produced by monocytes, macrophages, neutrophils, keratinocytes, fibroblasts, osteoclasts and endothelial cells, and is involved in inflammatory responses, tissue remodeling, wound healing, tumor growth and metastasis. MMP9 is supplied by bone marrow-derived cells and contributes to skin carcinogenesis. Further, MMP9 degrades type IV and V collagens. Studies in rhesus monkeys suggest that the enzyme is involved in IL-8-induced mobilization of hematopoietic progenitor cells from bone marrow, and murine studies suggest a role in tumor-associated tissue remodeling. Studies have shown that elevated MMP9 is associated with progression of idiopathic atrial fibrillation and aortic aneurysm.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: 92 kDa gelatinase; 92 kDa type IV collagenase; 92-kDa type IV collagenase; 92kD gelatinase; 92kD type IV collagenase; 92kDa gelatinase; 92kDa type IV collagenase; collagenase type IVB; Gel B; Gelatinase B; Gelatinase-B; GELB; macrophage gelatinase; matrix metallo protease; matrix metallopeptidase 9 (gelatinase B, 92kDa gelatinase, 92kDa type IV collagenase); matrix metalloproteinase 9; matrix metalloproteinase 9 (gelatinase B 92-kDa type IV collagenase); matrix metalloproteinase 9 (gelatinase B, 92-kDa type IV collagenase); matrix metalloproteinase 9 (gelatinase B, 92kDa gelatinase, 92kDa type IV collagenase); Matrix metalloproteinase-9; Matrix metalloproteinase9; MMP; MMP-9; MMP9; MMPs; type V collagenase; unnamed protein product

          View more View less

          Gene Aliases: AW743869; B/MMP9; CLG4B; GELB; MANDP2; MMP-9; MMP9; pro-MMP-9

          View more View less

          UniProt ID: (Human) P14780, (Rat) P50282, (Mouse) P41245

          View more View less

          Entrez Gene ID: (Human) 4318, (Rat) 81687, (Mouse) 17395

          View more View less

          Function(s)
          endopeptidase activity metalloendopeptidase activity serine-type endopeptidase activity protein binding collagen binding peptidase activity metallopeptidase activity zinc ion binding hydrolase activity identical protein binding metal ion binding fibronectin binding protein complex binding metalloprotease
          Process(es)
          skeletal system development positive regulation of protein phosphorylation proteolysis apoptotic process embryo implantation cell migration extracellular matrix disassembly extracellular matrix organization macrophage differentiation collagen catabolic process endodermal cell differentiation positive regulation of apoptotic process negative regulation of apoptotic process positive regulation of DNA binding positive regulation of epidermal growth factor receptor signaling pathway ephrin receptor signaling pathway regulation of multicellular organismal process positive regulation of keratinocyte migration cellular response to lipopolysaccharide cellular response to UV-A positive regulation of release of cytochrome c from mitochondria regulation of neuroinflammatory response negative regulation of cation transmembrane transport response to amyloid-beta positive regulation of vascular smooth muscle cell proliferation negative regulation of epithelial cell differentiation involved in kidney development negative regulation of intrinsic apoptotic signaling pathway ossification response to hypoxia kidney development positive regulation of leukocyte migration response to oxidative stress parturition aging response to nutrient response to radiation response to heat response to mechanical stimulus response to high light intensity response to organic substance response to organic cyclic compound transformation of host cell by virus positive regulation of synaptic plasticity response to estradiol response to lipopolysaccharide response to retinoic acid response to vitamin A response to tumor necrosis factor response to nicotine response to drug response to ethanol positive regulation of angiogenesis negative regulation of fibroblast proliferation tissue remodeling leukocyte migration protein oligomerization response to hyperoxia cellular response to iron(III) ion cellular response to cytokine stimulus cellular response to interleukin-1 cellular response to tumor necrosis factor cellular response to low-density lipoprotein particle stimulus cellular response to cell-matrix adhesion positive regulation of receptor binding negative regulation of cation channel activity negative regulation of cysteine-type endopeptidase activity involved in apoptotic signaling pathway
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline