Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MRELLELSCC HSCPFSSTAA AKCFAGLLNK HPAGQQLDEF LQLAVDKVEA GLGSGPCRSQ AFTLLLWVTK ALVLRYHPLS SCLTARLMGL LSDPELGPAA ADGFSLLMSD CTDVLTRAGH AEVRIMFRQR FFTDNVPALV QGFHAAPPDV KPNYLKGLSH VLNRLPKPVL LPELPTLLSL LLEALSCPDC VVQLSTLSCL QPLLLEAPQV MSLHVDTLVT KFLNLSSSPS MAVRIAALQC MHALTRLPTP VLLPYKPQVI RALAKSLDDK KRLVRKEAVS ARGEWFLLGS PGS
Key component of the cytosolic iron-sulfur protein assembly (CIA) complex, a multiprotein complex that mediates the incorporation of iron-sulfur cluster into apoproteins specifically involved in DNA metabolism and genomic integrity. In the CIA complex, MMS19 acts as an adapter between early-acting CIA components and a subset of cellular target iron-sulfur proteins such as ERCC2/XPD, FANCJ and RTEL1, thereby playing a key role in nucleotide excision repair (NER) and RNA polymerase II (POL II) transcription. As part of the mitotic spindle-associated MMXD complex, plays a role in chromosome segregation, probably by facilitating iron-sulfur cluster assembly into ERCC2/XPD. Indirectly acts as a transcriptional coactivator of estrogen receptor (ER), via its role in iron-sulfur insertion into some component of the TFIIH-machinery.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hMMS19; homolog of yeast MMS19; MET18 homolog; MMS19 cytosolic iron-sulfur assembly component; MMS19 nucleotide excision repair homolog; MMS19 nucleotide excision repair protein homolog; MMS19-like (MET18 homolog, S. cerevisiae); MMS19-like protein
Gene Aliases: hMMS19; MET18; MMS19; MMS19L
UniProt ID: (Human) Q96T76
Entrez Gene ID: (Human) 64210
Molecular Function: scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support