Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This gene encodes a nucleoid protein localized to the mitochondria inner membrane. The encoded protein affects regulation of insulin secretion, mitochondrial ATP production, and myogenesis through modulation of mitochondrial respiratory chain activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Breast cancer-associated protein SGA-81M; cytochrome B protein synthesis 6 homolog; Mitochondrial nucleoid factor 1; Mitochondrial protein M19; Ubiquinol-cytochrome c reductase complex assembly factor 2
Gene Aliases: bA6B20.2; C6orf125; Cbp6; M19; MNF1; UQCC2
UniProt ID: (Human) Q9BRT2
Entrez Gene ID: (Human) 84300
Molecular Function:
chaperone
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support