Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: PPFVGHCGA DCGWDRGEVC RVCQNKLFES IQEGKYEFPD KDWAHISSEA KDLISKLLVR DAKQRLSAAQ VLQHPWVQGQ APEKGLPTPQ VLQRNSSTMD LTLFAAEAIA LNRQLSQHEE NELAEEPEAL ADGLCSMKLS PPCKSRLARR RALAQAGRGE DRSPPTAL (299-465 aa encoded by BC002755)
The Map kinase-interacting kinases (MKNK1 and MKNK2) are activated by Map kinases, regulate protein synthesis by phosphroylation eIF4E on Ser209, and may contribute to tumorigenesis. Overexpression of Mnk2 results in increased phosphorylation of endogenous eIF-4E, showing that it can act as an eIF-4E kinase in vivo. Mnk2 may play a role in the response to environmental stress and cytokines. Expression of active mutants of MNK1 and MNK2 in 293 cells diminishes cap-dependent translation relative to cap-independent translation in a transient reporter assay. Human Mnk2 is homologous to murine Mnk2 (approximately 94% identical) and human Mnk1 (71% identical). In vitro phosphorylation studies show that Mnk2 is a significantly better substrate than Mnk1 for extracellular signal-regulated kinase 2 (Erk2), p38MAPKalpha, and p38MAPKbeta.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MAP kinase signal-integrating kinase 1; MAP kinase-interacting serine/threonine-protein kinase 1; MAPK signal-integrating kinase 1; mnk1; OTTHUMP00000009675; OTTHUMP00000009676
Gene Aliases: 2410048M24Rik; MKNK1; MNK1
UniProt ID: (Human) Q9BUB5, (Mouse) O08605, (Rat) Q4G050
Entrez Gene ID: (Human) 8569, (Mouse) 17346, (Rat) 500526
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support