Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: DALRGHFRS IRLRSRRLDC AACGERPTVT DLLDYEAFCG SSATDKCRSL QLLSPEERVS VTDYKRLLDS GAFHLLLDVR PQVEVDICRL PHALHIPLKH LERRDAESLK LLKEAIWEEK QGTQEGA (279-404 aa encoded by BC015939 )
Molybdenum cofactor (MoCo) is necessary for the function of all molybdoenzymes. One of the enzymes required for the biosynthesis of MoCo is molybdopterin synthase (MPT synthase). The protein encoded by this gene adenylates and activates MPT synthase. This gene contains no introns. A pseudogene of this gene is present on chromosome 14.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Adenylyltransferase and sulfurtransferase MOCS3; dJ914P20.3; MOCS3; Molybdenum cofactor synthesis protein 3; Molybdopterin synthase sulfurylase; MPT synthase sulfurylase; UBA4, ubiquitin-activating enzyme E1 homolog; ubiquitin-like modifier activating enzyme 4
Gene Aliases: MOCS3; UBA4
UniProt ID: (Human) O95396
Entrez Gene ID: (Human) 27304
Molecular Function:
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support