Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MASREEVLAL QAEVAQREEE LNSLKQKLAS ALLAEQEPQP ERLVPVSPLP PKAALSRDEI LRYSRQLVLP ELGVHGQLRL GTACVLIVGC GGLGCPLAQY LAAAGVGRLG LVDYDVVEMS NLARQVLHGE ALAGQAKAFS AAASLRRLNS AVECVPYTQA LTPATALDLV RRYDVVADCS DNVPTRYLVN DACVLAGRPL VSASALRFEG QITVYHYDGG PCYRCIFPQP PPAETVTNCA DGGVLGVVTG VLGCLQALEV LKIAAGLGPS YSGSLLLFDA LRGHFRSIRL RSRRLDCAAC GERPTVTDLL DYEAFCGSSA TDKCRSLQLL SPEERVSVTD YKRLLDSGAF HLLLDVRPQV EVDICRLPHA LHIPLKHLER RDAESLKLLK EAIWEEKQGT QEGAAVPIYV ICKLGNDSQK AVKILQSLSA AQELDPLTVR DVVGGLMAWA AKIDGTFPQY
Molybdenum cofactor (MoCo) is necessary for the function of all molybdoenzymes. One of the enzymes required for the biosynthesis of MoCo is molybdopterin synthase (MPT synthase). The protein encoded by this gene adenylates and activates MPT synthase. This gene contains no introns. A pseudogene of this gene is present on chromosome 14.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Adenylyltransferase and sulfurtransferase MOCS3; dJ914P20.3; MOCS3; Molybdenum cofactor synthesis protein 3; Molybdopterin synthase sulfurylase; MPT synthase sulfurylase; UBA4, ubiquitin-activating enzyme E1 homolog; ubiquitin-like modifier activating enzyme 4
Gene Aliases: MOCS3; UBA4
UniProt ID: (Human) O95396
Entrez Gene ID: (Human) 27304
Molecular Function:
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support