Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: YLVWLYVDWD TPNQGGRRSE WIRNRAIWRQ LRDYYPVKLV KTAELPPDR
Catalyzes the formation of diacylglycerol from 2- monoacylglycerol and fatty acyl-CoA. Also able to catalyze the terminal step in triacylglycerol synthesis by using diacylglycerol and fatty acyl-CoA as substrates. Has a preference toward palmitoyl-CoA and oleoyl-CoA. May be involved in absorption of dietary fat in the small intestine by catalyzing the resynthesis of triacylglycerol in enterocytes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2-acylglycerol O-acyltransferase 3; acyl coenzyme A:monoacylglycerol acyltransferase 3; Acyl-CoA:monoacylglycerol acyltransferase 3; Diacylglycerol acyltransferase 2-like protein 7; Diacylglycerol O-acyltransferase candidate 7; hDC7; MGAT3; Monoacylglycerol O-acyltransferase 3
Gene Aliases: DC7; DGAT2L2; DGAT2L7; MGAT3; MOGAT3; UNQ9383/PRO34208
UniProt ID: (Human) Q86VF5
Entrez Gene ID: (Human) 346606
Molecular Function: acyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support