Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bethyl Laboratories
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 2
The recommended shelf life for this product is 1 year from date of receipt.
May be required for traffic between late Golgi and early endosomes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MON2 regulator of endosome-to-Golgi trafficking; Protein MON2 homolog; Protein SF21
Gene Aliases: 2610528O22Rik; AW495628; KIAA1040; mKIAA1040; MON2; SF21
UniProt ID: (Human) Q7Z3U7, (Mouse) Q80TL7
Entrez Gene ID: (Human) 23041, (Mouse) 67074
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support