Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Osenses
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Reconstitute in 100 µL of sterile water. Centrifuge to remove any insoluble material.
Specificity of this antibody: MOR262 (subfamily 6, 3, 5, 7, 11, 12).
Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms. Subcellular Location: Cell membrane; Multi-pass membrane protein
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GA_x6K02T2N78B-16110014-16110970; GA_x6K02T2N78B-16125144-16126100; GA_x6K02T2N78B-16165641-16166600; GA_x6K02T2N78B-7084885-7085844; olfactory receptor 156; olfactory receptor 157; olfactory receptor 159; olfactory receptor 272; olfactory receptor 37b; olfactory receptor 37c; olfactory receptor 37e; olfactory receptor MOR262-12; olfactory receptor MOR262-5; olfactory receptor MOR262-6; olfactory receptor MOR262-7
Gene Aliases: MOR262-11; Mor262-12; MOR262-5; MOR262-6; MOR262-7; mOR37b; mOR37d; mOR37e; MTPCR52; Olfr158; Olfr29; Olfr37b; Olfr37c; Olfr37d; Olfr37e; OR37B; Or37c
Entrez Gene ID: (Mouse) 29846, (Mouse) 100040268, (Mouse) 29849, (Mouse) 258836, (Mouse) 29848
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support