Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 5
Adding 0.2 mL of distilled water will yield a concentration of 500 µg/mL.
This gene encodes a protein that localizes to the nuclear matrix. The protein also has RNA binding activity, and has a predicted coiled coil domain.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: microrchidia 3; MORC family CW-type zinc finger protein 3; Nuclear matrix protein 2; nuclear matrix protein NXP2; Zinc finger CW-type coiled-coil domain protein 3; zinc finger, CW type with coiled-coil domain 3; zinc finger, CW-type with coiled-coil domain 3
Gene Aliases: 1110051N18Rik; AI452146; BF318192; D16Jhu32e; KIAA0136; MORC3; NXP2; ZCW5; ZCWCC3
UniProt ID: (Human) Q14149, (Mouse) F7BJB9
Entrez Gene ID: (Human) 23515, (Rat) 304074, (Mouse) 338467
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support