Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide Sequence: MPARSGAQFC RRMGQKKQRP ARAGQPHSSS DAAQAPAEQP HSSSDAAQAP CPRERCLGPP TTPGPYRSIY FSSPKGHLTR LGLEFFDQPA
MPG functions in the hydrolysis of the deoxyribose N-glycosidic bond to excise 3-methyladenine, and 7-methylguanine from the damaged DNA polymer formed by alkylation lesions.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 3' end of the Mid1 gene, localized 68 kb upstream the humanzeta globin gene on 16p; 3-alkyladenine DNA glycosylase; 3-methyladenine DNA glycosidase; ADPG; CRA36.1 (3-methyl-adenine DNA glycosylase); DNA-3-methyladenine glycosylase; N-methylpurine-DNA glycosylase; N-methylpurine-DNA glycosylase, MPG; proliferation-inducing protein 11; proliferation-inducing protein 16
Gene Aliases: AAG; ADPG; ANPG; APNG; CRA36.1; MDG; MID1; MPG; PIG11; PIG16
UniProt ID: (Human) Q13770
Entrez Gene ID: (Human) 4350
Molecular Function:
DNA metabolism protein
DNA glycosylase
nucleic acid metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support