Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: PLGVTFRVE NNDLVIARIL HGGMIDRQGL LHVGDIIKEV NGHEVGNNPK ELQELLKNIS GSVTLKILPS YRDTITPQQV FVKCHFDYNP YNDNLIPCKE AGLKFSKGEI LQIVNREDPN WWQASHVKEG GSAGLIPSQF LEEKRKAFVR RDWDNSGPFC GTISSKKKKK MMYLTTRNAE FDRHEIQIYE EVAKMPPFQR KTLVLIGAQG VGRRSLKNRF IVLNPTRFGT TVPFTSRKPR EDEKDGQAYK FVSRSEMEAD IKAGKYLEHG EYEGNLYGTK IDSILEVVQT GRTCILDVNP QALKVLRTSE FMPYVVFIAA PELETLRAMH KAVVDAGITT KLLTDSDLKK TVDESARIQR AYNHYFDLII INDNLDKAFE KLQTAIEKLR MEPQWVPISW VY (140-540 aa encoded by BC023638)
Members of the peripheral membrane-associated guanylate kinase (MAGUK) family function in tumor suppression and receptor clustering by forming multiprotein complexes containing distinct sets of transmembrane, cytoskeletal, and cytoplasmic signaling proteins. All MAGUKs contain a PDZ-SH3-GUK core and are divided into 4 subfamilies, DLG-like (see DLG1), ZO1-like (see TJP1), p55-like (see MPP1), and LIN2-like (see CASK), based on their size and the presence of additional domains. MPP6 is a member of the p55-like MAGUK subfamily (Tseng et al., 2001).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CH474011:EDL88200; Dlgh4 protein; Fibroblast growth factor-inducible protein 15; MAGUK p55 subfamily member 6; MAGUK protein p55T; membrane protein, palmitoylated 3 (MAGUK p55 subfamily member 6); Membrane protein, palmitoylated 6; membrane protein, palmitoylated 6 (MAGUK p55 subfamily member 6); P55T protein; Protein associated with Lin-7 2; protein associated with Lin7 2; Protein PALS2; VAM-1; VELI-associated MAGUK 1
Gene Aliases: Dlgh4; MPP6; P55t; PALS2; VAM-1; VAM1
UniProt ID: (Human) Q9NZW5, (Mouse) Q9JLB0
Entrez Gene ID: (Human) 51678, (Rat) 362359, (Mouse) 56524
Molecular Function: scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support