Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Displays low metallophosphoesterase activity (in vitro). May play a role in the development of the nervous system.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 239FB; Fetal brain protein 239; Fetal brain protein 239 homolog; Metallophosphoesterase domain-containing protein 2; Metallophosphoesterase MPPED2; putative C11orf8 homolog
Gene Aliases: 239FB; 2700082O15Rik; AV354767; AW060960; C11orf8; C11orf8h; FAM1B; MPPED2
UniProt ID: (Human) Q15777, (Rat) B1WBP0, (Mouse) Q9CZJ0
Entrez Gene ID: (Human) 744, (Rat) 362185, (Mouse) 77015
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support