Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Antigen-presenting molecule specialized in presenting microbial vitamin B metabolites. Involved in the development and expansion of a small population of T-cells expressing an invariant T-cell receptor alpha chain called mucosal-associated invariant T- cells (MAIT). MAIT lymphocytes are preferentially located in the gut lamina propria and therefore may be involved in monitoring commensal flora or serve as a distress signal. Expression and MAIT cell recognition seem to be ligand-dependent.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Class I histocompatibility antigen-like protein; histocompatibility-2 complex class 1-like sequence; Major histocompatibility complex class I-related gene protein; major histocompatibility complex, class I-like sequence; MCH class I; MHC class I-like antigen MR-1; MHC class I-related gene protein; MHC class-I related-gene protein
Gene Aliases: H2ls; HLALS; MR1; Mr1a
UniProt ID: (Human) Q95460, (Mouse) Q8HWB0
Entrez Gene ID: (Human) 3140, (Mouse) 15064
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support