Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MANGTNASAP YYSYEYYLDY LDLIPVDEKK LKAHKHSIVI AFWVSLAAFV VLLFLILLYM SWSASPQMRN SPKHHQTCPW SHGLNLHLCI QKCLPCHREP LATSQAQASS VEPGSRTGPD QPLRQESSST LPLGGFQTHP TLLWELTLNG GPLVRSKPSE PPPGDRTSQL QS
MRAP encodes a melanocortin receptor-interacting protein. The encoded protein regulates trafficking and function of the melanocortin 2 receptor in the adrenal gland. The encoded protein can also modulate signaling of other melanocortin receptors. Mutations in this gene have been associated with familial glucocorticoid deficiency type 2.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: B27; Fat cell-specific low molecular weight protein; Fat tissue-specific low MW protein; Melanocortin-2 receptor accessory protein
Gene Aliases: B27; C21orf61; FALP; FGD2; GCCD2; MRAP
UniProt ID: (Human) Q8TCY5
Entrez Gene ID: (Human) 56246
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support