Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MTLEAIRYS RGSLQILDQL LLPKQSRYEA VGSVHQAWEA IRAMKVRGAP AIALVGCLSL AVELQAGAGG PGLAALVAFV RDKLSFLVTA RPTAVNMARA ARDLADVAAR EAEREGATEE AVRERVICCT EDMLEKDLRD NRSIGDLGAR HLLERVAPSG GKVTVLTHCN TGALATAGYG TALGVIRSLH SLGRLEHAFC TETRPYNQGA RLTAFELVYE QIPATLITDS MVAAAMAHRG VSAVVVGADR VVANGDTANK VGTYQLAIVA KHHGIPFYVA APSSSCDLRL ETGKEIIIEE RPGQELTDVN GVRIAAPGIG VWNPAFDVTP HDLITGGIIT ELGVFAPEEL RTALTTTISS RDGTLDGPQM (1-369 aa encoded by BC001703)
MGC3207 catalyzes the interconversion of methylthioribose-1-phosphate (MTR-1-P) into methylthioribulose-1-phosphate (MTRu-1-P). Independently from catalytic activity, MGC3207 promotes cell invasion in response to constitutive RhoA activation by promoting FAK tyrosine phosphorylation and stress fiber turnover.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: M1Pi; M1Pi {ECO:0000255|HAMAP-Rule:MF_03119}; Mediator of RhoA-dependent invasion; Methylthioribose-1-phosphate isomerase; methylthioribose-1-phosphate isomerase homolog; methylthioribose-1-phosphate isomerase {ECO:0000255|HAMAP-Rule:MF_03119}; MGC3207; MRI1; MTR-1-P isomerase; MTR-1-P isomerase {ECO:0000255|HAMAP-Rule:MF_03119}; S-methyl-5-thioribose-1-phosphate isomerase; S-methyl-5-thioribose-1-phosphate isomerase 1; S-methyl-5-thioribose-1-phosphate isomerase {ECO:0000255|HAMAP-Rule:MF_03119}; Translation initiation factor eIF-2B subunit alpha/beta/delta-like protein; translation initiation factor eIF-2B subunit alpha/beta/delta-like protein {ECO:0000255|HAMAP-Rule:MF_03119}
Gene Aliases: 2410018C20Rik; MRDI; MRI1; MTNA; RGD1307789; UNQ6390/PRO21135; Ypr118w
UniProt ID: (Human) Q9BV20, (Rat) Q5HZE4, (Mouse) Q9CQT1
Entrez Gene ID: (Human) 84245, (Rat) 288912, (Mouse) 67873
Molecular Function: isomerase metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support