Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This target displays homology in the following species: Cow: 92%; Dog: 93%; Horse: 92%; Human: 86%; Mouse: 100%; Pig: 85%; Rabbit: 77%; Rat: 100%; Yeast: 92%; Zebrafish: 77%
MGC3207 catalyzes the interconversion of methylthioribose-1-phosphate (MTR-1-P) into methylthioribulose-1-phosphate (MTRu-1-P). Independently from catalytic activity, MGC3207 promotes cell invasion in response to constitutive RhoA activation by promoting FAK tyrosine phosphorylation and stress fiber turnover.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: M1Pi; Methylthioribose-1-phosphate isomerase; MGC3207; MTR-1-P isomerase; S-methyl-5-thioribose-1-phosphate isomerase; Translation initiation factor eIF-2B subunit alpha/beta/delta-like protein
Gene Aliases: 2410018C20Rik; Mri1
UniProt ID: (Mouse) Q9CQT1
Entrez Gene ID: (Mouse) 67873
Molecular Function: isomerase metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support