Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 5
Positive test controls include: HeLa, HeLa Cytoplasm extracts, 293T. The target is usually found in the following locations: Membrane, Multi-pass membrane protein.
Immunogen sequence: SQEKVGIVGR TGAGKSSLIS ALFRLSEPEG KIWIDKILTT EIGLHDLRKK MSIIPQEPVL FTGTMRKNLD PFNEHTDEEL WNALQEVQLK ETIEDLPGKM DTELAESGSN FSVGQRQLVC LARAILRKNQ ILIIDEATAN VDPRTDELIQ KKIREKFAHC TVLTIAHRLN TIIDSDKIMV LDSGRLKEYD EPYVLLQNKE SLFYKMVQQL GKAEAAALTE TAKQVYFKRN YPHIGHTDHM VTNTSNGQPS TLTIFETAL
The protein encoded by this gene is a member of the superfamily of ATP-binding cassette transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies. This protein is a member of the MRP subfamily which is involved in multi-drug resistance. The specific function of this protein has not yet been determined; however, this protein may play a role in cellular detoxification as a pump for its substrate, organic anions. Alternative splicing results in multiple splice variants encoding different isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ABC-tranporter; ATP-binding cassette protein C4; ATP-binding cassette sub-family C member 4; ATP-binding cassette subfamily C member 4; ATP-binding cassette, sub-family C (CFTR/MRP), member 4; bA464I2.1 (ATP-binding cassette, sub-family C (CFTR/MRP), member 4); canalicular multispecific organic anion transporter (ABC superfamily); EST170205; MOAT-B; MRP/cMOAT-related ABC transporter; Multi-specific organic anion transporter B; Multidrug resistance-associated protein 4; RP11-74A12.1
Gene Aliases: ABCC4; D630049P08Rik; MOAT-B; MOATB; MRP4
UniProt ID: (Human) O15439, (Rat) F1M3J4, (Mouse) E9Q236
Entrez Gene ID: (Human) 10257, (Rat) 170924, (Mouse) 239273
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support