Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MARNVVYPL YRLGGPQLRV FRTNFFIQLV RPGVAQPEDT VQFRIPMEMT RVDLRNYLEG IYNVPVAAVR TRVQHGSNKR RDHRNVRIKK PDYKVAYVQL AHGQTFTFPD LFPEKDESPE GSAADDLYSM LEEERQQRQS SDPRRGGVPS WFGL (1-153 aa encoded by BC027710)
MRPL23 are mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. MRPL23 encodes a 39S subunit protein and is biallelically expressed, despite its location within a region of imprinted genes on chromosome 11.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 39S ribosomal protein L23, mitochondrial; FLJ45387; L23 mitochondrial-related protein; L23mt; Large ribosomal subunit protein uL23m; MRP-L23; Ribosomal protein L23-like; ribosomal protein related to L23 (mitochondrial)
Gene Aliases: L23MRP; MRPL23; RPL23; RPL23L
UniProt ID: (Human) Q16540
Entrez Gene ID: (Human) 6150
Molecular Function: ribosomal protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support