Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Predicted to react with Mouse and Rat samples.
MRPL23 are mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. MRPL23 encodes a 39S subunit protein and is biallelically expressed, despite its location within a region of imprinted genes on chromosome 11.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 39S ribosomal protein L23, mitochondrial; FLJ45387; L23 mitochondrial-related protein; L23mt; Large ribosomal subunit protein uL23m; MRP-L23; Ribosomal protein L23-like; ribosomal protein L23-related product homolog; ribosomal protein related to L23 (mitochondrial)
Gene Aliases: L23MRP; MRPL23; rL23MRP; RPL23; Rpl23-rs; RPL23L
UniProt ID: (Human) Q16540, (Mouse) O35972, (Rat) Q63750
Entrez Gene ID: (Human) 6150, (Mouse) 19935, (Rat) 64360
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support