Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 From June 2-13 take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: MANLFIRKM VNPLLYLSRH TVKPRALSTF LFGSIRGAAP VAVEPGAAVR SLLSPGLLPH LLPALGFKNK TVLKKRCKDC YLVKRRGRWY VYCKTHPRHK QRQM (1-103 aa encoded by BC020642)
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. MRPL36 encodes a 39S subunit protein. A pseudogene corresponding to this gene is found on chromosome 2p.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 39S ribosomal protein L36, mitochondrial; BRCA1-interacting protein 1; L36mt; Large ribosomal subunit protein bL36m; putative BRCA1-interacting protein
Gene Aliases: BRIP1; L36mt; MRP-L36; MRPL36; PRPL36; RPMJ
UniProt ID: (Human) Q9P0J6
Entrez Gene ID: (Human) 64979
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support