Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Immunogen sequence: MAALGTVLF TGVRRLHCSA AAWAGGQWRL QQGLAANPSG YGPLTELPDC SYADGRPAPP MKGQLRRKAE RETFARRVVL LSQEMDAGLQ AWQLRQQKLQ EEQRKQENAL KPKGASLKSP LPSQ (1-123 aa encoded by BC068070)
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein which has no bacterial homolog. Multiple transcript variants encoding different protein isoforms were identified through sequence analysis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 39S ribosomal protein L52, mitochondrial; L52mt; Large ribosomal subunit protein mL52; MRP-L52
Gene Aliases: 1110047B07Rik; MRPL52
UniProt ID: (Human) Q86TS9, (Mouse) Q9D0Y8
Entrez Gene ID: (Human) 122704, (Mouse) 68836, (Rat) 361037
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support