Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAAALARLG LRPVKQVRVQ FCPFEKNVES TRTFLQTVSS EKVRSTNLNC SVIADVRHDG SEPCVDVLFG DGHRLIMRGA HLTALEMLTA FASHIRARDA AGSGDKPGAD TGR (1-112 aa encoded by BC012163)
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. MRPL53 encodes a 39S subunit protein. A pseudogene corresponding to this gene is found on chromosome 1p.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 39S ribosomal protein L53, mitochondrial; L53mt; Large ribosomal subunit protein mL53; MRP-L53
Gene Aliases: 1110007K17Rik; L53MT; MRPL53
UniProt ID: (Human) Q96EL3, (Mouse) Q9D1H8
Entrez Gene ID: (Human) 116540, (Mouse) 68499, (Rat) 362388
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support