Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MLRVAWRTL SLIRTRAVTQ VLVPGLPGGG SAKFPFNQWG LQPRSLLLQA ARGYVVRKPA QSRLDDDPPP STLLKDYQNV PGIEKVDDVV KRLLSLEMAN KKEMLKIKQE QFMKKIVANP EDTRSLEARI IALSVKIRSY EEHLEKHRKD KAHKRYLLMS IDQRKKMLKN LRNTNYDVFE KICWGLGIEY TFPPLYYRRA HRRFVTKKAL CIRVFQETQK LKKRRRALKA AAAAQKQAKR RNPDSPAKAI PKTLKDSQ (1-257 aa encoded by BC031336)
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. MRPS15 encodes a 28S subunit protein that belongs to the ribosomal protein S15P family. The encoded protein is more than two times the size of its E. coli counterpart, with the 12S rRNA binding sites conserved. Between human and mouse, the encoded protein is the least conserved among small subunit ribosomal proteins. Pseudogenes corresponding to this gene are found on chromosomes 15q and 19q.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 28S ribosomal protein S15, mitochondrial; FLJ11564; MRP-S15; S15mt; Small ribosomal subunit protein uS15m
Gene Aliases: 1500003E24Rik; 2410002B11Rik; DC37; MPR-S15; Mprs15; MRPS15; RPMS15; S15mt
UniProt ID: (Human) P82914, (Mouse) Q9DC71, (Rat) Q5XI37
Entrez Gene ID: (Human) 64960, (Mouse) 66407, (Rat) 298517
Molecular Function: ribosomal protein translational protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support