Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: MAALKALVS GCGRLLRGLL AGPAATSWSR LPARGFREVV ETQEGKTTII EGRITATPKE SPNPPNPSGQ CPICRWNLKH KYNYDDVLLL SQFIRPHGGM LPRKITGLCQ EEHRKIEECV KMAHRAGLLP NHRPRLPEGV VPKSKPQLNR YLTRWAPGSV KPIYKKGPRW NRVRMPVGSP LLRDNVCYSR TPWKLYH
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 28S subunit protein that belongs to the ribosomal protein S18P family. The encoded protein is one of three that has significant sequence similarity to bacterial S18 proteins. The primary sequences of the three human mitochondrial S18 proteins are no more closely related to each other than they are to the prokaryotic S18 proteins. A pseudogene corresponding to this gene is found on chromosome 3p. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 28S ribosomal protein S18-3, mitochondrial; 39S ribosomal protein S18-3, mitochondrial; 39S ribosomal protein S18a, mitochondrial; Large ribosomal subunit protein bS18a; Large ribosomal subunit protein mL66; mitochondrial ribosomal protein S18-3; MRP-S18-3; MRP-S18-a; S18mt-a
Gene Aliases: HumanS18b; MRP-S18-3; MRPS18-3; MRPS18A; S18bmt
UniProt ID: (Human) Q9NVS2
Entrez Gene ID: (Human) 55168
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support