Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAAVVAVCGG LGRKKLTHLV TAAVSLTHPG THTVLWRRGC SQQVSSNEDL PISMENPYKE PLKKCILCGK HVDYKNVQLL SQFVSPFTGC IYGRHITGLC GKKQKEITKA IKRAQIMGFM PVTYKDPAYL KDPKVCNIRY RE
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 28S subunit protein that belongs to the ribosomal protein S18P family. The encoded protein is one of three that has significant sequence similarity to bacterial S18 proteins. The primary sequences of the three human mitochondrial S18 proteins are no more closely related to each other than they are to the prokaryotic S18 proteins. Pseudogenes corresponding to this gene are found on chromosomes 8p, 12p, 15q, and 22q. [provided by RefSeq, Jul 2008].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 28S ribosomal protein S18-1, mitochondrial; 28S ribosomal protein S18c, mitochondrial; mitochondrial ribosomal protein S18-1; MRP-S18-1; MRP-S18-c; Small ribosomal subunit protein bS18c; Small ribosomal subunit protein bS18m
Gene Aliases: CGI-134; MRP-S18-1; MRP-S18-c; MRPS18-1; mrps18-c; MRPS18C; S18mt-c
UniProt ID: (Human) Q9Y3D5
Entrez Gene ID: (Human) 51023
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support