Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: YLDRALQVME KVAASPEDIK LCREALDVLG AVLKALTSAD GASEEQSQND EDNQGSEKLV EQLDIEETEQ SKLPQYLERF KALHSKLQAL GKIESEGLLS LTTQLVKEKL STCEAEDIAT YEQNLQQWHL DLVQLIQREQ QQREQAKQEY QAQKAAKASA; Positive Samples: A-549, LO2, U-87MG, HeLa, A-431, HT-29, Mouse liver, Rat heart; Cellular Location: Mitochondrion
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 28S subunit protein that may be a functional partner of the death associated protein 3 (DAP3).
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 28S ribosomal protein S27, mitochondrial; mitochondrial 28S ribosomal protein S27; Mitochondrial ribosomal protein S27; MRP-S27; mrps27; S27mt; Small ribosomal subunit protein mS27
Gene Aliases: 2610028H14Rik; KIAA0264; MRP-S27; MRPS27; S27mt
UniProt ID: (Human) Q92552, (Mouse) Q8BK72
Entrez Gene ID: (Human) 23107, (Mouse) 218506, (Rat) 361883
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support