Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MARKKVRPR LIAELARRVR ALREQLNRPR DSQLYAVDYE TLTRPFSGRR LPVRAWADVR RESRLLQLLG RLPLFGLGRL VTRKSWLWQH DEPCYWRLTR VRPDYTAQNL DHGKAWGILT FKGKTESEAR EIEHVMYHDW RLVPKHEEEA FTAFTPAPED SLASVPYPPL LRAMIIAERQ KNGDTSTEEP MLNVQRIRME PWDYPAKQED KGRAKGTPV (1-218 aa encoded by BC001182 )
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 28S subunit protein. Alternate splice variants of this gene have been described but their full-length nature has not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 28S ribosomal protein S34, mitochondrial; mitochondrial 28S ribosomal protein S34; MRP-S34; rt34; S34mt; Small ribosomal subunit protein mS34; T-complex expressed gene 2 protein
Gene Aliases: 0610007F04Rik; 5330430D13Rik; AV001970; MRP-S12; MRP-S34; MRPS12; MRPS34; Tce2
UniProt ID: (Human) P82930, (Mouse) Q9JIK9
Entrez Gene ID: (Human) 65993, (Rat) 287126, (Mouse) 79044
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support