Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Immunogen sequence: SSGYVEPMQ TLNDVLAQLD AVVSFAHVSN GAPVPYVRPA ILEKGQGRII LKASRHACVE VQDEIAFIPN DVYFEKDKQM FHIITGPNMG GKSTYIRQTG VIVLMAQIGC FVPCESAEVS IVDCILARVG AGDSQLKGVS TFMAEMLETA SILRSATKDS LIIIDELGRG TSTYDGFGLA WAISEYIATK IGAFCMFATH FHELTALANQ IPTVNNLHVT ALTTEETLTM LYQVKKGVCD QSFGIHVAEL ANFPKHVIEC AKQKALELEE FQYIGESQGY DIMEPAAKKC YLEREQGEKI IQEFLSKVKQ MPFTEMSEEN ITIKLKQLKA EVIAKNNSFV NEIISRIKVT T (585-934 aa encoded by BC021566)
Plays a role in neurogenesis and neuronal migration. Necessary for correct formation of mitotic spindles and chromosome separation during mitosis. Necessary for cytokinesis and cell proliferation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DNA mismatch repair protein Msh2; hMSH2; Msh2p; mutS homolog 2, colon cancer, nonpolyposis type 1; MutS protein homolog 2
Gene Aliases: COCA1; FCC1; HNPCC; HNPCC1; LCFS2; MSH2
UniProt ID: (Human) P43246
Entrez Gene ID: (Human) 4436
Molecular Function: DNA metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support