Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
MSK1 is a mitogen and stress activated protein kinase 1 which belongs to the AGC family of kinases and is related in structure to the ribosomal p70 S6 kinase subfamily. MSK1 can be activated by ERK1/2 and SAPK2/p38 MAP kinase. It is also known to be required for the phosphorylation of CREB, ATF1 H3 and HMG14 in response to mitogen and stress. Similar to RSK, MSK1 contains two kinase domains (N term and a C term). Once phosphorylated on Thr581 and Ser360 by ERK1/2 and SAPK2/p38, MSK1 autophosphorylate on at least 5 sites. Of these autophosphorylation sites Ser212 and Ser376 get phosphorylated by the C terminal kinase domain of MSK1 which is essential for the catalytic activity of the N terminal kinase domain. MSK2 plays an essential role in the control of RELA transcriptional activity in response to TNF. Phosphorylates 'Ser-10' of histone H3 in response to mitogenics, stress stimuli and EGF, which results in the transcriptional activation of several immediate early genes, including proto-oncogenes c-fos/FOS and c-jun/JUN. May also phosphorylate 'Ser-28' of histone H3. Mediates the mitogen- and stress-induced phosphorylation of high mobility group protein 1 (HMGN1/HMG14). In lipopolysaccharide-stimulated primary macrophages, acts downstream of the Toll-like receptor TLR4 to limit the production of pro-inflammatory cytokines. Functions probably by inducing transcription of the MAP kinase phosphatase DUSP1 and the anti-inflammatory cytokine interleukin 10 (IL10), via CREB1 and ATF1 transcription factors.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 90 kDa ribosomal protein S6 kinase 4; 90 kDa ribosomal protein S6 kinase 5; coiled-coil domain containing 88B; mitogen and stress-activated protein kinase 1; mitogen- and stress-activated protein kinase 2; mitogen- and stress-activated protein kinase-2; Nuclear mitogen- and stress-activated protein kinase 1; Nuclear mitogen- and stress-activated protein kinase 2; Ribosomal protein kinase B; ribosomal protein S6 kinase alpha 4; Ribosomal protein S6 kinase alpha-4; Ribosomal protein S6 kinase alpha-5; ribosomal protein S6 kinase, 90kD, polypeptide 4; ribosomal protein S6 kinase, 90kDa, polypeptide 4; ribosomal protein S6 kinase, 90kDa, polypeptide 5; RLSK; RSK-like protein kinase; RSKB; RSKL; S6K-alpha-4; S6K-alpha-5
Gene Aliases: 1110069D02Rik; 3110005L17Rik; 6330404E13Rik; 90kDa; AI848992; AI854034; Ccdc88b; mMSK2; MSK1; MSK2; MSPK1; RLPK; RPS6KA4; RPS6KA5; RSK-B; S6K-alpha-4
UniProt ID: (Human) O75676, (Human) O75582, (Mouse) Q9Z2B9, (Mouse) Q8C050
Entrez Gene ID: (Human) 8986, (Human) 9252, (Mouse) 56613, (Rat) 361715, (Mouse) 73086, (Rat) 314384
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support