Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen Sequence: LRKRLTPLQY HVTQEKGTES AFEGEYTHHK DPGIYKCVVC GTPLFKSETK FDSGSGWPSF HDVINSEAIT FTDDFSYGMH RVETSCSQCG AHLGHIFDDG PRPTGKRYCI NSAALSFTPA DSSGTAEGGS GVASPAQADK AEL
The protein encoded by this gene catalyzes the reduction of methionine sulfoxide to methionine. This enzyme acts as a monomer and requires zinc as a cofactor. Several transcript variants encoding two different isoforms have been found for this gene. One of the isoforms localizes to mitochondria while the other localizes to endoplasmic reticula.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: methionine sulfoxide reductase B3; Methionine-R-sulfoxide reductase B3; Methionine-R-sulfoxide reductase B3, mitochondrial; MsrB3
Gene Aliases: D430026P16Rik; DFNB74; I-8-23; MsrB2; MSRB3; UNQ1965/PRO4487
UniProt ID: (Human) Q8IXL7, (Mouse) Q8BU85
Entrez Gene ID: (Human) 253827, (Mouse) 320183, (Rat) 680036
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support