Save up to 31% on select cell culture essentials

Bundle and save

Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            Target of Interest
            Application
            Target Species
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • YBX2 Antibodies

            Proteintech

            MSY2 Polyclonal Antibody

            6 References
            View all (20) YBX2 antibodies
            Datasheet
            Protocols
            Questions & Answers
            Catalog # 13538-1-AP
            Web quote
            You can generate a web quote (PDF) by signing in to your account and adding products to your cart. View cart and click the 'Generate a web quote' button. With your web quote document, you can get approval for your order and complete your order online.
             
            Product not available in your region
            150 µL
             
            Join Aspire™ for free antibodies
            Datasheet
            Protocols
            Questions & Answers

            Cite MSY2 Polyclonal Antibody

            • Antibody Testing Data (6)
            Application
            MSY2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            Group 53 Created with Sketch.

            FIGURE: 1 / 6

            MSY2 Antibody (13538-1-AP) in IHC (P)

            Immunohistochemistry of paraffin-embedded human ovary tumor using 13538-1-AP (MSY2 antibody) at dilution of 1:50 (under 10x lens).
            View Product

            MSY2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            MSY2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            MSY2 Antibody in Western Blot (WB)
            MSY2 Antibody in Western Blot (WB)
            MSY2 Antibody in Western Blot (WB)
            MSY2 Antibody in Immunoprecipitation (IP)

            Product Details

            13538-1-AP

            Applications
            Tested Dilution
            Publications

            Western Blot (WB)

            1:500-1:1,000
            View 2 publications 2 publications

            Immunohistochemistry (IHC)

            -
            View 1 publication 1 publication

            Immunohistochemistry (Paraffin) (IHC (P))

            1:20-1:200
            -

            Immunocytochemistry (ICC/IF)

            -
            View 1 publication 1 publication

            Immunoprecipitation (IP)

            0.5-4.0 µg
            View 1 publication 1 publication

            ChIP assay (ChIP)

            -
            View 1 publication 1 publication
            Product Specifications

            Species Reactivity

            Human, Mouse, Rat

            Host/Isotype

            Rabbit / IgG

            Class

            Polyclonal

            Type

            Antibody

            Immunogen

            MSY2 Fusion Protein Ag4465 (1-364 aa encoded by BC033800)
            View immunogen

            Conjugate

            Unconjugated

            Form

            Liquid

            Concentration

            0.33 mg/mL

            Purification

            Antigen affinity chromatography

            Storage buffer

            PBS, pH 7.3, with 50% glycerol

            Contains

            0.02% sodium azide

            Storage conditions

            -20°C

            Shipping conditions

            Wet ice (domestic); Dry ice (international)

            Product Specific Information

            Immunogen sequence: MSEVEAAAG ATAVPAATVP ATAAGVVAVV VPVPAGEPQK GGGAGGGGGA ASGPAAGTPS APGSRTPGNP ATAVSGTPAP PARSQADKPV LAIQVLGTVK WFNVRNGYGF INRNDTKEDV FVHQTAIKRN NPRKFLRSVG DGETVEFDVV EGEKGAEATN VTGPGGVPVK GSRYAPNRRK SRRFIPRPPS VAPPPMVAEI PSAGTGPGSK GERAEDSGQR PRRWCPPPFF YRRRFVRGPR PPNQQQPIEG TDRVEPKETA PLEGHQQQGD ERVPPPRFRP RYRRPFRPRP RQQPTTEGGD GETKPSQGPA DGSRPEPQRP RNRPYFQRRR QQAPGPQQAP GPRQPAAPET SAPVNSGDPT TTILE (1-364 aa encoded by BC033800)

            Target Information

            YBX2 is a germ-cell specific member of the vertebrate Y box family of nucleic acid-binding proteins.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            Bioinformatics

            Protein Aliases: Contrin; Dbpc; DNA-binding protein C; FRGY2 homolog; germ cell specific Y-box binding protein; Germ cell-specific Y-box-binding protein; MSY2 homolog; Y box binding protein 2; Y box protein 2; Y-box protein MSY2; Y-box-binding protein 2

            View more View less

            Gene Aliases: CONTRIN; CSDA3; DBPC; MSY2; RGD1305068; YBX2

            View more View less

            UniProt ID: (Human) Q9Y2T7

            View more View less

            Entrez Gene ID: (Human) 51087, (Mouse) 53422, (Rat) 303250

            View more View less

            Function(s)

            DNA binding RNA binding nucleic acid binding chromatin binding mRNA binding mRNA 3'-UTR binding lipid binding ribonucleoprotein complex binding translation regulator activity molecular_function

            Process(es)

            regulation of transcription, DNA-templated transcription from RNA polymerase II promoter spermatogenesis translational attenuation oocyte development spermatid development negative regulation of translation mRNA stabilization oogenesis negative regulation of binding
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            united states flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-56756dfb8b-mfpfg:80/100.66.79.199:80.
            git-commit: de6905f249d25e22729e613aef254bb544bd8b7c
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.31.1-2025.06.25-1.0