Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide sequence: DGVVVVVNFN SVGSWMIYEG EGSGLIREDP IGAGALYDYG RWLVVVTGWT
Sequence homology: Human: 100%; Pig: 93%
Core subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I) that is believed to belong to the minimal assembly required for catalysis. Complex I functions in the transfer of electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: NADH dehydrogenase subunit 6; NADH-ubiquinone oxidoreductase chain 6
Gene Aliases: MT-ND6; MTND6; NADH6; ND6
UniProt ID: (Human) P03923
Entrez Gene ID: (Human) 4541
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support