Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Positive Samples: Mouse liver, Mouse heart, Mouse kidney, Rat liver, Rat kidney; Cellular Location: Mitochondrion inner membrane, Multi-pass membrane protein
Immunogen sequence: GHQWYWSYEY TDYEDLCFDS YMIPTNDLKP GELRLLEVDN RVVLPMELPI RMLISSEDVL HSWAVPSLGL KTDAIPGRLN QATVTSNRPG LFYGQCSEIC
Component of the cytochrome c oxidase, the last enzyme in the mitochondrial electron transport chain which drives oxidative phosphorylation. The respiratory chain contains 3 multisubunit complexes succinate dehydrogenase (complex II, CII), ubiquinol-cytochrome c oxidoreductase (cytochrome b-c1 complex, complex III, CIII) and cytochrome c oxidase (complex IV, CIV), that cooperate to transfer electrons derived from NADH and succinate to molecular oxygen, creating an electrochemical gradient over the inner membrane that drives transmembrane transport and the ATP synthase. Cytochrome c oxidase is the component of the respiratory chain that catalyzes the reduction of oxygen to water. Electrons originating from reduced cytochrome c in the intermembrane space (IMS) are transferred via the dinuclear copper A center (CU(A)) of subunit 2 and heme A of subunit 1 to the active site in subunit 1, a binuclear center (BNC) formed by heme A3 and copper B (CU(B)). The BNC reduces molecular oxygen to 2 water molecules using 4 electrons from cytochrome c in the IMS and 4 protons from the mitochondrial matrix. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Co2; cyclooxygenase; Cytochrome c oxidase polypeptide II; Cytochrome c oxidase subunit 2
Gene Aliases: COII; COX2; COXII; MT-CO2; MTCO2
UniProt ID: (Mouse) P00405
Entrez Gene ID: (Mouse) 17709
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support